Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D12
Entrez Gene ID
23232
Gene Name
TBC1 domain family member 12
Gene Aliases
-
Chromosome Location
Chr.10: 94402429 - 94536332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015188.1 NP_056003.1 8 - 9 1873 135
XM_006717732.3 XP_006717795.2 7 - 8 1792 135
XM_006717733.2 XP_006717796.1 9 - 10 833 135
XM_011539558.2 XP_011537860.1 8 - 9 884 135
XM_017015996.1 XP_016871485.1 6 - 7 1648 135
XM_017015998.1 XP_016871487.1 7 - 8 660 135
XM_017015999.1 XP_016871488.1 7 - 8 876 135
GenBank mRNA AB011180.1 - 8 - 9 1689 135
AB449898.1 - 8 - 9 1670 135
AK090865.1 - 1 - 2 1740 135
AL049956.1 - 7 - 8 621 135

View Full Product Details