The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142633.2 | NP_001136105.1 | 17 - 18 | 2514 | 64 |
NM_001251851.1 | NP_001238780.1 | 17 - 18 | 2576 | 64 | |
NM_001251852.1 | NP_001238781.1 | 16 - 17 | 2398 | 64 | |
NM_001251853.1 | NP_001238782.1 | 16 - 17 | 2360 | 64 | |
NM_001251855.1 | NP_001238784.1 | 16 - 17 | 2339 | 64 | |
NM_014308.3 | NP_055123.2 | 17 - 18 | 2451 | 64 | |
XM_005256580.3 | XP_005256637.1 | 17 - 18 | 2506 | 64 | |
GenBank mRNA | AB028925.1 | - | 16 - 17 | 2384 | 64 |
AF128881.1 | - | 17 - 18 | 2451 | 64 | |
AK314186.1 | - | 16 - 17 | 2581 | 64 | |
BC028212.1 | - | 16 - 17 | 2330 | 64 |