Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOLT1B
Entrez Gene ID
51026
Gene Name
golgi transport 1B
Gene Aliases
CGI-141, GCT2, GOT1, GOT1B, YMR292W
Chromosome Location
Chr.12: 21501765 - 21518403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK310889.1 - 3 - 4 379 80

View Full Product Details