Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELL
Entrez Gene ID
6402
Gene Name
selectin L
Gene Aliases
CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1
Chromosome Location
Chr.1: 169690665 - 169711702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000655.4 NP_000646.2 6 - 7 1150 113
NR_029467.1 4 - 5 914 113
GenBank mRNA AJ246000.1 - 6 - 7 1021 113
AK225713.1 - 6 - 7 1061 113
AK298705.1 - 4 - 5 909 113
AK312673.1 - 6 - 7 1061 113
AY367061.1 - 4 - 5 514 113
BC020758.1 - 6 - 7 1047 113
M25280.1 - 6 - 7 1061 113
X16070.1 - 6 - 7 1074 113
X16150.1 - 6 - 7 1037 113
X17519.1 - 6 - 7 1034 113

View Full Product Details