The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018429.2 | NP_060899.2 | 24 - 25 | 5652 | 127 |
XM_011543511.2 | XP_011541813.1 | 24 - 25 | 5630 | 127 | |
XM_011543512.2 | XP_011541814.1 | 24 - 25 | 5631 | 127 | |
XM_017009630.1 | XP_016865119.1 | 24 - 25 | 5631 | 127 | |
XM_017009631.1 | XP_016865120.1 | 24 - 25 | 5631 | 127 | |
XM_017009632.1 | XP_016865121.1 | 24 - 25 | 5630 | 127 | |
XM_017009633.1 | XP_016865122.1 | 24 - 25 | 5630 | 127 | |
XM_017009634.1 | XP_016865123.1 | 24 - 25 | 5630 | 127 | |
GenBank mRNA | AB051476.3 | - | 24 - 25 | 5399 | 127 |
AJ238520.2 | - | 24 - 25 | 5593 | 127 | |
BC146792.1 | - | 24 - 25 | 5399 | 127 | |
BC161615.1 | - | 24 - 25 | 5381 | 127 |