Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location
Chr.10: 114431110 - 114821643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003407.1 NP_001003407.1 16 - 17 1952 84
NM_001003408.1 NP_001003408.1 17 - 18 2036 84
NM_001322882.1 NP_001309811.1 17 - 18 2042 84
NM_001322883.1 NP_001309812.1 15 - 16 1847 84
NM_001322884.1 NP_001309813.1 15 - 16 1652 84
NM_001322885.1 NP_001309814.1 15 - 16 1646 84
NM_001322886.1 NP_001309815.1 15 - 16 1610 84
NM_001322887.1 NP_001309816.1 14 - 15 1706 84
NM_001322888.1 NP_001309817.1 11 - 12 1182 84
NM_001322889.1 NP_001309818.1 11 - 12 1167 84
NM_001322890.1 NP_001309819.1 10 - 11 1104 84
NM_001322891.1 NP_001309820.1 10 - 11 1125 84
NM_001322892.1 NP_001309821.1 10 - 11 1098 84
NM_001322893.1 NP_001309822.1 10 - 11 1119 84
NM_001322894.1 NP_001309823.1 10 - 11 1083 84
NM_001322895.1 NP_001309824.1 10 - 11 1062 84
NM_001322896.1 NP_001309825.1 9 - 10 1020 84
NM_001322897.1 NP_001309826.1 9 - 10 984 84
NM_001322898.1 NP_001309827.1 9 - 10 957 84
NM_001322899.1 NP_001309828.1 9 - 10 978 84
NM_001322900.1 NP_001309829.1 8 - 9 879 84
NM_002313.5 NP_002304.3 16 - 17 1929 84
NM_006720.3 NP_006711.3 11 - 12 1182 84
XM_005269824.2 XP_005269881.1 15 - 16 2547 84
XM_005269826.1 XP_005269883.1 13 - 14 1418 84
XM_005269827.4 XP_005269884.1 12 - 13 2048 84
XM_006717837.1 XP_006717900.1 18 - 19 2510 84
XM_006717846.3 XP_006717909.1 13 - 14 2153 84
XM_006717847.1 XP_006717910.1 12 - 13 1247 84
XM_011539801.1 XP_011538103.1 18 - 19 1914 84
XM_011539802.1 XP_011538104.1 18 - 19 1827 84
XM_011539803.1 XP_011538105.1 18 - 19 1806 84
XM_011539805.1 XP_011538107.1 18 - 19 2167 84
XM_017016242.1 XP_016871731.1 20 - 21 2010 84
XM_017016243.1 XP_016871732.1 19 - 20 1896 84
XM_017016244.1 XP_016871733.1 19 - 20 1890 84
XM_017016245.1 XP_016871734.1 18 - 19 2000 84
XM_017016246.1 XP_016871735.1 18 - 19 1791 84
XM_017016247.1 XP_016871736.1 15 - 16 1565 84
XM_017016248.1 XP_016871737.1 15 - 16 1565 84
XM_017016249.1 XP_016871738.1 18 - 19 1749 84
XM_017016250.1 XP_016871739.1 17 - 18 1916 84
XM_017016251.1 XP_016871740.1 17 - 18 1707 84
XM_017016252.1 XP_016871741.1 17 - 18 1701 84
XM_017016253.1 XP_016871742.1 17 - 18 1665 84
XM_017016254.1 XP_016871743.1 17 - 18 1644 84
XM_017016255.1 XP_016871744.1 16 - 17 1796 84
XM_017016256.1 XP_016871745.1 15 - 16 2589 84
XM_017016258.1 XP_016871747.1 11 - 12 1928 84
XM_017016259.1 XP_016871748.1 11 - 12 1127 84
GenBank mRNA AF005654.1 - 16 - 17 1950 84
AK058007.1 - 6 - 7 571 84
AK093471.1 - 16 - 17 1451 84
AK098277.1 - 11 - 12 1187 84
AK122891.1 - 17 - 18 2392 84
AK297344.1 - 15 - 16 1885 84
AK298709.1 - 9 - 10 861 84
BC002448.2 - 9 - 10 924 84
CR749819.1 - 10 - 11 1094 84
D31883.2 - 16 - 17 1754 84

View Full Product Details