Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101802.1 NP_001095272.1 12 - 13 1846 75
NM_016621.3 NP_057705.3 12 - 13 1849 75
XM_005252965.4 XP_005253022.1 13 - 14 1538 75
XM_011520157.2 XP_011518459.1 12 - 13 1399 75
XM_011520158.2 XP_011518460.1 13 - 14 1751 75
XM_011520160.2 XP_011518462.1 15 - 16 1912 75
XM_011520161.2 XP_011518463.1 15 - 16 2003 75
XM_011520162.2 XP_011518464.1 15 - 16 1881 75
XM_011520164.2 XP_011518466.1 14 - 15 1810 75
XM_011520165.2 XP_011518467.1 14 - 15 1765 75
XM_011520166.2 XP_011518468.1 14 - 15 1688 75
XM_011520167.2 XP_011518469.1 14 - 15 1891 75
XM_011520168.2 XP_011518470.1 14 - 15 1743 75
XM_011520169.2 XP_011518471.1 13 - 14 1702 75
XM_011520173.2 XP_011518475.1 13 - 14 1658 75
XM_011520174.2 XP_011518476.1 14 - 15 1632 75
XM_011520175.2 XP_011518477.1 14 - 15 1629 75
XM_011520179.2 XP_011518481.1 13 - 14 1652 75
XM_017017885.1 XP_016873374.1 15 - 16 1831 75
XM_017017886.1 XP_016873375.1 14 - 15 1796 75
XM_017017887.1 XP_016873376.1 13 - 14 1576 75
XM_017017888.1 XP_016873377.1 13 - 14 1748 75
XM_017017889.1 XP_016873378.1 13 - 14 1748 75
XM_017017890.1 XP_016873379.1 13 - 14 1745 75
XM_017017891.1 XP_016873380.1 13 - 14 1655 75
XM_017017892.1 XP_016873381.1 13 - 14 1655 75
XM_017017893.1 XP_016873382.1 13 - 14 1751 75
XM_017017894.1 XP_016873383.1 13 - 14 1751 75
XM_017017895.1 XP_016873384.1 13 - 14 1748 75
XM_017017896.1 XP_016873385.1 13 - 14 1748 75
XM_017017897.1 XP_016873386.1 14 - 15 1828 75
XM_017017898.1 XP_016873387.1 14 - 15 1686 75
XM_017017899.1 XP_016873388.1 15 - 16 1820 75
XM_017017900.1 XP_016873389.1 14 - 15 1775 75
XM_017017901.1 XP_016873390.1 13 - 14 1751 75
XM_017017902.1 XP_016873391.1 13 - 14 1655 75
XM_017017903.1 XP_016873392.1 10 - 11 1305 75
XM_017017904.1 XP_016873393.1 13 - 14 1751 75
XM_017017905.1 XP_016873394.1 12 - 13 1499 75
XM_017017906.1 XP_016873395.1 10 - 11 1305 75
XM_017017907.1 XP_016873396.1 12 - 13 1406 75
XM_017017908.1 XP_016873397.1 15 - 16 1919 75
GenBank mRNA AB051483.1 - 12 - 13 1593 75
AF208848.1 - 4 - 5 380 75
AK021530.1 - 12 - 13 1497 75
AK023258.1 - 14 - 15 1610 75
AY090777.1 - 10 - 11 1225 75
BC015714.2 - 13 - 14 1510 75
BX648236.1 - 5 - 6 2080 75

View Full Product Details