Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101802.1 NP_001095272.1 16 - 17 2304 127
NM_016621.3 NP_057705.3 16 - 17 2166 127
XM_005252965.4 XP_005253022.1 17 - 18 1996 127
XM_011520157.2 XP_011518459.1 16 - 17 1857 127
XM_011520158.2 XP_011518460.1 17 - 18 2209 127
XM_011520160.2 XP_011518462.1 19 - 20 2370 127
XM_011520161.2 XP_011518463.1 19 - 20 2461 127
XM_011520162.2 XP_011518464.1 19 - 20 2339 127
XM_011520164.2 XP_011518466.1 18 - 19 2268 127
XM_011520165.2 XP_011518467.1 18 - 19 2223 127
XM_011520166.2 XP_011518468.1 18 - 19 2146 127
XM_011520167.2 XP_011518469.1 18 - 19 2349 127
XM_011520168.2 XP_011518470.1 18 - 19 2201 127
XM_011520169.2 XP_011518471.1 17 - 18 2160 127
XM_011520173.2 XP_011518475.1 17 - 18 2116 127
XM_011520174.2 XP_011518476.1 18 - 19 2090 127
XM_011520175.2 XP_011518477.1 18 - 19 2087 127
XM_011520179.2 XP_011518481.1 17 - 18 1969 127
XM_017017885.1 XP_016873374.1 19 - 20 2289 127
XM_017017886.1 XP_016873375.1 18 - 19 2254 127
XM_017017887.1 XP_016873376.1 17 - 18 2034 127
XM_017017888.1 XP_016873377.1 17 - 18 2206 127
XM_017017889.1 XP_016873378.1 17 - 18 2206 127
XM_017017890.1 XP_016873379.1 17 - 18 2203 127
XM_017017891.1 XP_016873380.1 17 - 18 2113 127
XM_017017892.1 XP_016873381.1 17 - 18 2113 127
XM_017017893.1 XP_016873382.1 17 - 18 2209 127
XM_017017894.1 XP_016873383.1 17 - 18 2068 127
XM_017017895.1 XP_016873384.1 17 - 18 2065 127
XM_017017896.1 XP_016873385.1 17 - 18 2065 127
XM_017017897.1 XP_016873386.1 18 - 19 2286 127
XM_017017898.1 XP_016873387.1 18 - 19 2144 127
XM_017017899.1 XP_016873388.1 19 - 20 2278 127
XM_017017900.1 XP_016873389.1 18 - 19 2233 127
XM_017017901.1 XP_016873390.1 16 - 17 2053 127
XM_017017902.1 XP_016873391.1 17 - 18 1972 127
XM_017017903.1 XP_016873392.1 14 - 15 1622 127
XM_017017904.1 XP_016873393.1 17 - 18 2209 127
XM_017017905.1 XP_016873394.1 16 - 17 1957 127
XM_017017906.1 XP_016873395.1 14 - 15 1622 127
XM_017017907.1 XP_016873396.1 16 - 17 1864 127
XM_017017908.1 XP_016873397.1 19 - 20 2377 127
GenBank mRNA AB051483.1 - 16 - 17 1910 127
AF208848.1 - 8 - 9 697 127
AK023258.1 - 18 - 19 1927 127
AY090777.1 - 14 - 15 1542 127
BC015714.2 - 17 - 18 1968 127
BX648236.1 - 9 - 10 2538 127

View Full Product Details