Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016621.3 NP_057705.3 7 - 8 1237 66
XM_005252965.4 XP_005253022.1 8 - 9 926 66
XM_011520157.2 XP_011518459.1 7 - 8 787 66
XM_011520158.2 XP_011518460.1 8 - 9 1139 66
XM_011520160.2 XP_011518462.1 10 - 11 1300 66
XM_011520161.2 XP_011518463.1 10 - 11 1391 66
XM_011520162.2 XP_011518464.1 10 - 11 1269 66
XM_011520164.2 XP_011518466.1 9 - 10 1198 66
XM_011520165.2 XP_011518467.1 9 - 10 1153 66
XM_011520166.2 XP_011518468.1 9 - 10 1076 66
XM_011520167.2 XP_011518469.1 9 - 10 1279 66
XM_011520168.2 XP_011518470.1 9 - 10 1131 66
XM_011520169.2 XP_011518471.1 8 - 9 1090 66
XM_011520173.2 XP_011518475.1 8 - 9 1046 66
XM_011520174.2 XP_011518476.1 9 - 10 1020 66
XM_011520175.2 XP_011518477.1 9 - 10 1017 66
XM_011520179.2 XP_011518481.1 8 - 9 1040 66
XM_017017885.1 XP_016873374.1 10 - 11 1219 66
XM_017017886.1 XP_016873375.1 9 - 10 1184 66
XM_017017887.1 XP_016873376.1 8 - 9 964 66
XM_017017889.1 XP_016873378.1 8 - 9 1136 66
XM_017017892.1 XP_016873381.1 8 - 9 1043 66
XM_017017893.1 XP_016873382.1 8 - 9 1139 66
XM_017017894.1 XP_016873383.1 8 - 9 1139 66
XM_017017896.1 XP_016873385.1 8 - 9 1136 66
XM_017017897.1 XP_016873386.1 9 - 10 1216 66
XM_017017898.1 XP_016873387.1 9 - 10 1074 66
XM_017017899.1 XP_016873388.1 10 - 11 1208 66
XM_017017900.1 XP_016873389.1 9 - 10 1163 66
XM_017017901.1 XP_016873390.1 8 - 9 1139 66
XM_017017902.1 XP_016873391.1 8 - 9 1043 66
XM_017017904.1 XP_016873393.1 8 - 9 1139 66
XM_017017906.1 XP_016873395.1 5 - 6 693 66
GenBank mRNA AB051483.1 - 7 - 8 981 66
AK021530.1 - 7 - 8 885 66
AK023258.1 - 9 - 10 998 66
AY090777.1 - 5 - 6 613 66

View Full Product Details