Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307943.1 NP_001294872.1 11 - 12 1550 113
NM_018259.5 NP_060729.2 11 - 12 1550 113
XM_005253016.1 XP_005253073.1 11 - 12 1550 113
XM_006718263.1 XP_006718326.1 9 - 10 1233 113
XM_011520218.2 XP_011518520.1 11 - 12 1550 113
XM_017018010.1 XP_016873499.1 11 - 12 1550 113
GenBank mRNA AK001540.1 - 6 - 7 785 113
AK001752.1 - 11 - 12 1503 113
AK074253.1 - 1 - 2 67 113
BC017238.2 - 11 - 12 1499 113
BC033000.1 - 11 - 12 1496 113
BX640972.1 - 6 - 7 1390 113

View Full Product Details