The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307943.1 | NP_001294872.1 | 16 - 17 | 2320 | 89 |
XM_005253016.1 | XP_005253073.1 | 16 - 17 | 2320 | 89 | |
XM_006718263.1 | XP_006718326.1 | 14 - 15 | 2003 | 89 | |
XM_011520218.2 | XP_011518520.1 | 16 - 17 | 2320 | 89 | |
XM_017018010.1 | XP_016873499.1 | 16 - 17 | 2320 | 89 | |
GenBank mRNA | AK001540.1 | - | 11 - 12 | 1555 | 89 |
AK001752.1 | - | 16 - 17 | 2273 | 89 | |
BC033000.1 | - | 16 - 17 | 2266 | 89 | |
BX640972.1 | - | 11 - 12 | 2160 | 89 |