The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307943.1 | NP_001294872.1 | 17 - 18 | 2497 | 68 |
XM_005253016.1 | XP_005253073.1 | 17 - 18 | 2497 | 68 | |
XM_006718263.1 | XP_006718326.1 | 15 - 16 | 2180 | 68 | |
XM_011520218.2 | XP_011518520.1 | 17 - 18 | 2497 | 68 | |
XM_017018010.1 | XP_016873499.1 | 17 - 18 | 2497 | 68 | |
GenBank mRNA | AF161431.1 | - | 1 - 2 | 97 | 68 |
AK001540.1 | - | 12 - 13 | 1732 | 68 | |
AK001752.1 | - | 17 - 18 | 2450 | 68 | |
AK023161.1 | - | 1 - 2 | 126 | 68 | |
BC033000.1 | - | 17 - 18 | 2443 | 68 | |
BX640972.1 | - | 12 - 13 | 2337 | 68 |