Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018259.5 NP_060729.2 18 - 19 2730 56
XM_005253016.1 XP_005253073.1 19 - 20 2901 56
XM_006718263.1 XP_006718326.1 17 - 18 2584 56
XM_011520218.2 XP_011518520.1 19 - 20 2901 56
GenBank mRNA AK023161.1 - 3 - 4 530 56
AK074253.1 - 8 - 9 1247 56
BC017238.2 - 18 - 19 2679 56

View Full Product Details