Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018259.5 NP_060729.2 23 - 24 3363 69
XM_005253016.1 XP_005253073.1 24 - 25 3534 69
XM_006718263.1 XP_006718326.1 22 - 23 3217 69
GenBank mRNA AK023161.1 - 8 - 9 1163 69
AK026217.1 - 1 - 2 1058 69
AK074253.1 - 13 - 14 1880 69
AK124380.1 - 4 - 5 1991 69
BC017238.2 - 23 - 24 3312 69
BC041893.1 - 1 - 2 1445 69

View Full Product Details