Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307943.1 NP_001294872.1 7 - 8 991 95
NM_018259.5 NP_060729.2 7 - 8 991 95
XM_005253016.1 XP_005253073.1 7 - 8 991 95
XM_006718263.1 XP_006718326.1 5 - 6 674 95
XM_011520218.2 XP_011518520.1 7 - 8 991 95
XM_017018010.1 XP_016873499.1 7 - 8 991 95
GenBank mRNA AK001540.1 - 2 - 3 226 95
AK001752.1 - 7 - 8 944 95
BC017238.2 - 7 - 8 940 95
BC033000.1 - 7 - 8 937 95
BX640972.1 - 2 - 3 831 95

View Full Product Details