Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307943.1 NP_001294872.1 8 - 9 1133 112
NM_018259.5 NP_060729.2 8 - 9 1133 112
XM_005253016.1 XP_005253073.1 8 - 9 1133 112
XM_006718263.1 XP_006718326.1 6 - 7 816 112
XM_011520218.2 XP_011518520.1 8 - 9 1133 112
XM_017018010.1 XP_016873499.1 8 - 9 1133 112
GenBank mRNA AK001540.1 - 3 - 4 368 112
AK001752.1 - 8 - 9 1086 112
BC017238.2 - 8 - 9 1082 112
BC033000.1 - 8 - 9 1079 112
BX640972.1 - 3 - 4 973 112

View Full Product Details