The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077186.1 | NP_001070654.1 | 27 - 28 | 3626 | 65 |
NM_001145809.1 | NP_001139281.1 | 28 - 29 | 3725 | 65 | |
NM_024729.3 | NP_079005.3 | 26 - 27 | 3602 | 65 | |
XM_006723386.3 | XP_006723449.1 | 27 - 28 | 3687 | 65 | |
XM_011527320.2 | XP_011525622.1 | 27 - 28 | 3704 | 65 | |
XM_011527321.2 | XP_011525623.1 | 26 - 27 | 3680 | 65 | |
XM_011527323.2 | XP_011525625.1 | 27 - 28 | 3694 | 65 | |
GenBank mRNA | AB111886.1 | - | 17 - 18 | 2284 | 65 |
AB290169.1 | - | 28 - 29 | 3695 | 65 | |
AY165122.1 | - | 27 - 28 | 3602 | 65 | |
AY203926.1 | - | 5 - 6 | 667 | 65 | |
BC129803.1 | - | 15 - 16 | 1699 | 65 |