The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077186.1 | NP_001070654.1 | 28 - 29 | 3775 | 133 |
NM_001145809.1 | NP_001139281.1 | 29 - 30 | 3874 | 133 | |
NM_024729.3 | NP_079005.3 | 27 - 28 | 3751 | 133 | |
XM_006723386.3 | XP_006723449.1 | 28 - 29 | 3836 | 133 | |
XM_011527320.2 | XP_011525622.1 | 28 - 29 | 3853 | 133 | |
XM_011527321.2 | XP_011525623.1 | 27 - 28 | 3829 | 133 | |
XM_011527323.2 | XP_011525625.1 | 28 - 29 | 3843 | 133 | |
GenBank mRNA | AB111886.1 | - | 18 - 19 | 2433 | 133 |
AB290169.1 | - | 29 - 30 | 3844 | 133 | |
AY165122.1 | - | 28 - 29 | 3751 | 133 | |
AY203926.1 | - | 6 - 7 | 816 | 133 | |
BC129803.1 | - | 16 - 17 | 1848 | 133 |