The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077186.1 | NP_001070654.1 | 29 - 30 | 3975 | 59 |
NM_001145809.1 | NP_001139281.1 | 30 - 31 | 4074 | 59 | |
NM_024729.3 | NP_079005.3 | 28 - 29 | 3951 | 59 | |
XM_006723386.3 | XP_006723449.1 | 29 - 30 | 4036 | 59 | |
XM_011527320.2 | XP_011525622.1 | 29 - 30 | 4053 | 59 | |
XM_011527321.2 | XP_011525623.1 | 28 - 29 | 4029 | 59 | |
XM_011527323.2 | XP_011525625.1 | 29 - 30 | 4043 | 59 | |
GenBank mRNA | AB111886.1 | - | 19 - 20 | 2633 | 59 |
AB290169.1 | - | 30 - 31 | 4044 | 59 | |
AY165122.1 | - | 29 - 30 | 3951 | 59 | |
AY203926.1 | - | 7 - 8 | 1016 | 59 | |
BC129803.1 | - | 17 - 18 | 2048 | 59 |