Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYH14
Entrez Gene ID
79784
Gene Name
myosin, heavy chain 14, non-muscle
Gene Aliases
DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
Chromosome Location
Chr.19: 50188186 - 50310544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077186.1 NP_001070654.1 33 - 34 4701 103
NM_001145809.1 NP_001139281.1 34 - 35 4800 103
NM_024729.3 NP_079005.3 32 - 33 4677 103
XM_006723386.3 XP_006723449.1 33 - 34 4762 103
XM_011527320.2 XP_011525622.1 33 - 34 4779 103
XM_011527321.2 XP_011525623.1 32 - 33 4755 103
XM_011527323.2 XP_011525625.1 33 - 34 4769 103
GenBank mRNA AB111886.1 - 23 - 24 3359 103
AB290169.1 - 34 - 35 4770 103
AK023943.1 - 2 - 3 341 103
AK122704.1 - 1 - 2 1928 103
AK303704.1 - 3 - 4 627 103
AY165122.1 - 33 - 34 4677 103
AY203926.1 - 11 - 12 1741 103
BC129803.1 - 21 - 22 2774 103
CR936653.1 - 3 - 4 629 103

View Full Product Details