The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077186.1 | NP_001070654.1 | 37 - 38 | 5296 | 62 |
NM_001145809.1 | NP_001139281.1 | 38 - 39 | 5395 | 62 | |
NM_024729.3 | NP_079005.3 | 36 - 37 | 5272 | 62 | |
XM_006723386.3 | XP_006723449.1 | 37 - 38 | 5357 | 62 | |
XM_011527320.2 | XP_011525622.1 | 37 - 38 | 5374 | 62 | |
XM_011527321.2 | XP_011525623.1 | 36 - 37 | 5350 | 62 | |
XM_011527323.2 | XP_011525625.1 | 37 - 38 | 5364 | 62 | |
GenBank mRNA | AB111886.1 | - | 27 - 28 | 3954 | 62 |
AB290169.1 | - | 38 - 39 | 5365 | 62 | |
AK023943.1 | - | 6 - 7 | 936 | 62 | |
AK122704.1 | - | 5 - 6 | 2523 | 62 | |
AK303704.1 | - | 7 - 8 | 1222 | 62 | |
AY165122.1 | - | 37 - 38 | 5272 | 62 | |
AY203926.1 | - | 15 - 16 | 2336 | 62 | |
BC000676.2 | - | 7 - 8 | 690 | 62 | |
BC129803.1 | - | 25 - 26 | 3369 | 62 | |
CR936653.1 | - | 7 - 8 | 1224 | 62 | |
DB110776.1 | - | 3 - 4 | 345 | 62 |