Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYH14
Entrez Gene ID
79784
Gene Name
myosin, heavy chain 14, non-muscle
Gene Aliases
DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
Chromosome Location
Chr.19: 50188186 - 50310544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077186.1 NP_001070654.1 40 - 41 5736 107
NM_001145809.1 NP_001139281.1 41 - 42 5835 107
NM_024729.3 NP_079005.3 39 - 40 5712 107
XM_006723386.3 XP_006723449.1 40 - 41 5797 107
XM_011527320.2 XP_011525622.1 40 - 41 5814 107
XM_011527321.2 XP_011525623.1 39 - 40 5790 107
XM_011527323.2 XP_011525625.1 40 - 41 5804 107
GenBank mRNA AB111886.1 - 30 - 31 4394 107
AB290169.1 - 41 - 42 5805 107
AK023943.1 - 9 - 10 1376 107
AK122704.1 - 8 - 9 2963 107
AK303704.1 - 10 - 11 1662 107
AY165122.1 - 40 - 41 5712 107
AY203926.1 - 18 - 19 2777 107
BC000676.2 - 10 - 11 1130 107
BC007877.2 - 3 - 4 450 107
BC129803.1 - 28 - 29 3809 107
CR936653.1 - 10 - 11 1664 107

View Full Product Details