The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308384.1 | NP_001295313.1 | 6 - 7 | 1636 | 60 |
NM_018231.2 | NP_060701.1 | 7 - 8 | 1168 | 60 | |
XM_017023393.1 | XP_016878882.1 | 6 - 7 | 1649 | 60 | |
XM_017023394.1 | XP_016878883.1 | 7 - 8 | 1144 | 60 | |
XM_017023395.1 | XP_016878884.1 | 7 - 8 | 1144 | 60 | |
XM_017023396.1 | XP_016878885.1 | 7 - 8 | 1144 | 60 | |
XM_017023397.1 | XP_016878886.1 | 7 - 8 | 1144 | 60 | |
XM_017023398.1 | XP_016878887.1 | 7 - 8 | 1144 | 60 | |
GenBank mRNA | AK001677.1 | - | 7 - 8 | 1161 | 60 |
AK022786.1 | - | 7 - 8 | 1134 | 60 | |
AK222932.1 | - | 7 - 8 | 1130 | 60 | |
BC001961.1 | - | 6 - 7 | 1117 | 60 | |
BC063399.1 | - | 6 - 7 | 1636 | 60 | |
CR457236.1 | - | 5 - 6 | 768 | 60 |