Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TXNDC5
- Entrez Gene ID
-
81567
- Gene Name
- thioredoxin domain containing 5
- Gene Aliases
-
ENDOPDI, ERP46, HCC-2, HCC2, PDIA15, STRF8, UNQ364
- Chromosome Location
-
Chr.6: 7881250 - 7910814 on Build GRCh38
- Gene Symbol
- BLOC1S5-TXNDC5
- Entrez Gene ID
-
100526836
- Gene Name
- BLOC1S5-TXNDC5 readthrough (NMD candidate)
- Gene Aliases
-
MUTED-TXNDC5
- Chromosome Location
-
Chr.6: 7881250 - 8064414 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037616.1 |
|
5 - 6 |
577 |
138 |
GenBank mRNA |
BC052310.1 |
- |
5 - 6 |
527 |
138 |
View Full Product Details