Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS16
Entrez Gene ID
170690
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 16
Gene Aliases
ADAMTS16s
Chromosome Location
Chr.5: 5140330 - 5320304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139056.3 NP_620687.2 1 - 2 212 150
NR_136935.1 1 - 2 212 150
GenBank mRNA AJ315734.1 - 1 - 2 74 150
AK127746.1 - 1 - 2 212 150
DQ266047.1 - 1 - 2 212 150

View Full Product Details