Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG4B
Entrez Gene ID
23192
Gene Name
autophagy related 4B cysteine peptidase
Gene Aliases
APG4B, AUTL1
Chromosome Location
Chr.2: 241637612 - 241673857 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005246995.3 XP_005247052.1 2 - 3 194 138
XM_005246996.2 XP_005247053.1 2 - 3 139 138
XM_017003639.1 XP_016859128.1 1 - 2 122 229
XM_017003640.1 XP_016859129.1 2 - 3 128 229
GenBank mRNA AK027332.1 - 1 - 2 72 138
AK296818.1 - 2 - 3 139 138
BX648007.1 - 2 - 3 138 138

View Full Product Details