The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178044.1 | NP_001171515.1 | 16 - 17 | 1701 | 101 |
NM_001178045.1 | NP_001171516.1 | 17 - 18 | 1830 | 101 | |
NM_025257.2 | NP_079533.2 | 17 - 18 | 1827 | 101 | |
GenBank mRNA | AK027397.1 | - | 17 - 18 | 1778 | 101 |
AK222998.1 | - | 17 - 18 | 1778 | 101 | |
AK300550.1 | - | 17 - 18 | 1830 | 101 | |
AK301596.1 | - | 16 - 17 | 1652 | 101 | |
AK314673.1 | - | 17 - 18 | 1782 | 101 | |
AL833009.1 | - | 13 - 14 | 1520 | 101 | |
AY358457.1 | - | 5 - 6 | 594 | 101 | |
BC014659.1 | - | 17 - 18 | 1766 | 101 | |
KU175229.1 | - | 17 - 18 | 1761 | 101 |