Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
Chromosome Location
Chr.2: 85354394 - 85391752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135023.1 NP_001128495.1 2 - 3 204 111
XM_005264597.3 XP_005264654.1 2 - 3 530 111
GenBank mRNA BC112324.1 - 2 - 3 167 111

View Full Product Details