Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR75
Entrez Gene ID
84128
Gene Name
WD repeat domain 75
Gene Aliases
NET16, UTP17
Chromosome Location
Chr.2: 189441433 - 189475562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303096.1 NP_001290025.1 20 - 21 2288 117
NM_032168.2 NP_115544.1 19 - 20 2254 117
GenBank mRNA AK022581.1 - 19 - 20 2201 117
AK023276.1 - 16 - 17 1893 117
AK091546.1 - 20 - 21 2215 117
AK290445.1 - 19 - 20 2205 117
AL122097.1 - 15 - 16 1821 117
BC006816.1 - 4 - 5 448 117
BC018689.1 - 2 - 3 413 117
BC040567.1 - 19 - 20 2254 117
BU619995.1 - 2 - 3 117
CR749440.1 - 19 - 20 3863 117

View Full Product Details