The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013338.4 | NP_037470.1 | 4 - 5 | 415 | 121 |
XM_017020552.1 | XP_016876041.1 | 3 - 4 | 353 | 121 | |
GenBank mRNA | AF102850.1 | - | 4 - 5 | 423 | 121 |
AF161498.1 | - | 4 - 5 | 364 | 121 | |
AF183413.1 | - | 4 - 5 | 374 | 121 | |
AK299126.1 | - | 4 - 5 | 384 | 121 | |
BC012531.1 | - | 4 - 5 | 367 | 121 | |
BF668393.1 | - | - | 389 | 121 | |
BG482604.1 | - | 4 - 5 | 400 | 121 | |
BP246765.1 | - | 4 - 5 | 415 | 121 | |
CB988638.1 | - | - | 412 | 121 |