The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014243.2 | NP_055058.2 | 6 - 7 | 978 | 69 |
XM_011532421.2 | XP_011530723.1 | 6 - 7 | 1010 | 69 | |
XM_011532422.2 | XP_011530724.1 | 6 - 7 | 1509 | 69 | |
GenBank mRNA | AB002364.1 | - | 6 - 7 | 930 | 69 |
BC130287.1 | - | 6 - 7 | 975 | 69 | |
BC132735.1 | - | 6 - 7 | 975 | 69 |