The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174908.3 | NP_777568.1 | 3 - 4 | 832 | 80 |
NM_178335.2 | NP_848018.1 | 3 - 4 | 832 | 80 | |
XM_011512460.1 | XP_011510762.1 | 3 - 4 | 835 | 80 | |
GenBank mRNA | AJ416916.2 | - | 3 - 4 | 832 | 80 |
AJ557013.1 | - | 3 - 4 | 832 | 80 | |
BC065004.1 | - | 3 - 4 | 329 | 80 |