The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098484.2 | NP_001091954.1 | 2 - 3 | 190 | 81 |
NM_001134742.1 | NP_001128214.1 | 2 - 3 | 190 | 81 | |
GenBank mRNA | AB470072.1 | - | 1 - 2 | 80 | 81 |
AF011390.1 | - | 2 - 3 | 190 | 81 | |
AF053753.1 | - | 1 - 2 | 73 | 81 | |
AF053754.1 | - | 1 - 2 | 73 | 81 | |
AF069510.1 | - | 1 - 2 | 117 | 81 | |
AF157492.1 | - | 2 - 3 | 149 | 81 | |
AF310248.1 | - | 1 - 2 | 73 | 81 | |
EF531618.1 | - | 1 - 2 | 73 | 81 |