Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
Chromosome Location
Chr.16: 2425438 - 2458858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001761.2 NP_001752.2 2 - 3 260 69
XM_017023822.1 XP_016879311.1 2 - 3 420 69
XM_017023823.1 XP_016879312.1 2 - 3 420 69
GenBank mRNA AK313371.1 - 2 - 3 213 69
BC012349.1 - 2 - 3 184 69
HY027359.1 - 2 - 3 263 69
U17105.1 - 2 - 3 185 69
Z36714.1 - 2 - 3 215 69

View Full Product Details