The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015060.2 | NP_055875.1 | 12 - 13 | 1856 | 132 |
XM_005249668.2 | XP_005249725.1 | 12 - 13 | 1805 | 132 | |
XM_005249669.2 | XP_005249726.1 | 12 - 13 | 1805 | 132 | |
XM_005249670.2 | XP_005249727.1 | 12 - 13 | 1511 | 132 | |
XM_017011891.1 | XP_016867380.1 | 12 - 13 | 1499 | 132 | |
XM_017011892.1 | XP_016867381.1 | 12 - 13 | 1509 | 132 | |
GenBank mRNA | AK090619.1 | - | 12 - 13 | 132 | |
CN312870.1 | - | 2 - 3 | 380 | 132 |