The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015060.2 | NP_055875.1 | 7 - 8 | 853 | 113 |
XM_005249668.2 | XP_005249725.1 | 7 - 8 | 790 | 113 | |
XM_005249669.2 | XP_005249726.1 | 7 - 8 | 790 | 113 | |
XM_005249670.2 | XP_005249727.1 | 7 - 8 | 790 | 113 | |
XM_005249671.4 | XP_005249728.1 | 7 - 8 | 790 | 113 | |
XM_011515227.2 | XP_011513529.1 | 7 - 8 | 790 | 113 | |
XM_017011891.1 | XP_016867380.1 | 7 - 8 | 790 | 113 | |
XM_017011892.1 | XP_016867381.1 | 7 - 8 | 800 | 113 | |
GenBank mRNA | AK090619.1 | - | 7 - 8 | 715 | 113 |
BC027724.2 | - | 7 - 8 | 705 | 113 | |
D87682.1 | - | 5 - 6 | 362 | 113 |