Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLTCL1
Entrez Gene ID
8218
Gene Name
clathrin heavy chain like 1
Gene Aliases
CHC22, CLH22, CLTCL, CLTD
Chromosome Location
Chr.22: 19179473 - 19291716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001835.3 NP_001826.3 11 - 12 1855 85
NM_007098.3 NP_009029.3 11 - 12 1855 85
XM_017028953.1 XP_016884442.1 11 - 12 1848 85
XM_017028954.1 XP_016884443.1 10 - 11 1734 85
XM_017028955.1 XP_016884444.1 11 - 12 1848 85
XM_017028956.1 XP_016884445.1 11 - 12 1848 85
XM_017028957.1 XP_016884446.1 11 - 12 1848 85
GenBank mRNA AK302506.1 - 11 - 12 1824 85
BC113030.1 - 4 - 5 551 85
BC143977.1 - 11 - 12 1800 85
BC143979.1 - 11 - 12 1800 85
U41763.1 - 11 - 12 1877 85
U60802.1 - 11 - 12 1824 85
X95486.1 - 11 - 12 1813 85

View Full Product Details