Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLTCL1
Entrez Gene ID
8218
Gene Name
clathrin heavy chain like 1
Gene Aliases
CHC22, CLH22, CLTCL, CLTD
Chromosome Location
Chr.22: 19179473 - 19291716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001835.3 NP_001826.3 12 - 13 2025 61
NM_007098.3 NP_009029.3 12 - 13 2025 61
XM_011530401.2 XP_011528703.1 2 - 3 259 61
XM_017028953.1 XP_016884442.1 12 - 13 2018 61
XM_017028954.1 XP_016884443.1 11 - 12 1904 61
XM_017028955.1 XP_016884444.1 12 - 13 2018 61
XM_017028956.1 XP_016884445.1 12 - 13 2018 61
XM_017028957.1 XP_016884446.1 12 - 13 2018 61
GenBank mRNA AK302506.1 - 12 - 13 1994 61
BC143977.1 - 12 - 13 1970 61
BC143979.1 - 12 - 13 1970 61
U41763.1 - 12 - 13 2047 61
U60802.1 - 12 - 13 1994 61
X95486.1 - 12 - 13 1983 61

View Full Product Details