The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001835.3 | NP_001826.3 | 4 - 5 | 759 | 72 |
NM_007098.3 | NP_009029.3 | 4 - 5 | 759 | 72 | |
XM_017028953.1 | XP_016884442.1 | 4 - 5 | 752 | 72 | |
XM_017028955.1 | XP_016884444.1 | 4 - 5 | 752 | 72 | |
XM_017028956.1 | XP_016884445.1 | 4 - 5 | 752 | 72 | |
XM_017028957.1 | XP_016884446.1 | 4 - 5 | 752 | 72 | |
GenBank mRNA | AK302506.1 | - | 4 - 5 | 728 | 72 |
BC143977.1 | - | 4 - 5 | 704 | 72 | |
BC143979.1 | - | 4 - 5 | 704 | 72 | |
U60802.1 | - | 4 - 5 | 728 | 72 | |
X95486.1 | - | 4 - 5 | 717 | 72 |