The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032236.5 | NP_115612.4 | 18 - 19 | 2509 | 80 |
XM_005246009.2 | XP_005246066.1 | 18 - 19 | 2513 | 80 | |
XM_006710951.2 | XP_006711014.2 | 18 - 19 | 2507 | 80 | |
XM_006710955.3 | XP_006711018.1 | 17 - 18 | 2291 | 80 | |
XM_011542262.2 | XP_011540564.1 | 18 - 19 | 2403 | 80 | |
XM_011542263.2 | XP_011540565.1 | 18 - 19 | 2403 | 80 | |
XM_011542264.1 | XP_011540566.1 | 17 - 18 | 2103 | 80 | |
XM_011542265.2 | XP_011540567.1 | 19 - 20 | 3069 | 80 | |
XM_011542267.2 | XP_011540569.1 | 18 - 19 | 2422 | 80 | |
XM_011542268.2 | XP_011540570.1 | 10 - 11 | 1122 | 80 | |
XM_017002495.1 | XP_016857984.1 | 17 - 18 | 2312 | 80 | |
XM_017002497.1 | XP_016857986.1 | 10 - 11 | 1167 | 80 | |
GenBank mRNA | AF502942.1 | - | 18 - 19 | 2411 | 80 |
AK002190.1 | - | 2 - 3 | 140 | 80 | |
AK026707.1 | - | 7 - 8 | 1173 | 80 | |
AK226150.1 | - | 17 - 18 | 2307 | 80 | |
AK315005.1 | - | 18 - 19 | 2289 | 80 | |
AL834375.1 | - | 3 - 4 | 202 | 80 | |
BC013567.1 | - | 2 - 3 | 187 | 80 | |
BC104896.1 | - | 18 - 19 | 2305 | 80 | |
BC143343.1 | - | 18 - 19 | 2305 | 80 | |
BC143349.1 | - | 18 - 19 | 2305 | 80 | |
BC143354.1 | - | 18 - 19 | 2341 | 80 |