The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032236.5 | NP_115612.4 | 22 - 23 | 2966 | 80 |
XM_006710951.2 | XP_006711014.2 | 22 - 23 | 2964 | 80 | |
XM_006710955.3 | XP_006711018.1 | 21 - 22 | 2748 | 80 | |
XM_011542262.2 | XP_011540564.1 | 22 - 23 | 2860 | 80 | |
XM_011542263.2 | XP_011540565.1 | 22 - 23 | 2860 | 80 | |
XM_011542264.1 | XP_011540566.1 | 21 - 22 | 2560 | 80 | |
XM_011542265.2 | XP_011540567.1 | 23 - 24 | 3526 | 80 | |
XM_011542267.2 | XP_011540569.1 | 22 - 23 | 2879 | 80 | |
XM_011542268.2 | XP_011540570.1 | 14 - 15 | 1579 | 80 | |
XM_017002495.1 | XP_016857984.1 | 21 - 22 | 2769 | 80 | |
XM_017002497.1 | XP_016857986.1 | 14 - 15 | 1624 | 80 | |
GenBank mRNA | AF502942.1 | - | 22 - 23 | 2868 | 80 |
AK000110.1 | - | 2 - 3 | 460 | 80 | |
AK002190.1 | - | 6 - 7 | 597 | 80 | |
AK226150.1 | - | 21 - 22 | 2764 | 80 | |
AK315005.1 | - | 22 - 23 | 2746 | 80 | |
AL834375.1 | - | 7 - 8 | 659 | 80 | |
BC013567.1 | - | 6 - 7 | 644 | 80 | |
BC143354.1 | - | 22 - 23 | 2798 | 80 |