The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143778.1 | NP_001137250.1 | 10 - 11 | 1114 | 60 |
NM_017707.3 | NP_060177.2 | 11 - 12 | 1141 | 60 | |
XM_011541755.1 | XP_011540057.1 | 11 - 12 | 1081 | 60 | |
XM_017001685.1 | XP_016857174.1 | 11 - 12 | 1075 | 60 | |
XM_017001686.1 | XP_016857175.1 | 11 - 12 | 1081 | 60 | |
XM_017001687.1 | XP_016857176.1 | 11 - 12 | 1048 | 60 | |
XM_017001688.1 | XP_016857177.1 | 11 - 12 | 1037 | 60 | |
XM_017001689.1 | XP_016857178.1 | 11 - 12 | 1098 | 60 | |
GenBank mRNA | AB056749.1 | - | 11 - 12 | 1062 | 60 |
AK000253.1 | - | 1 - 2 | 60 | ||
AK092291.1 | - | 11 - 12 | 1063 | 60 | |
AK295100.1 | - | 10 - 11 | 1036 | 60 | |
AK299359.1 | - | 11 - 12 | 1037 | 60 | |
BC060786.1 | - | 11 - 12 | 1089 | 60 |