The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152443.2 | NP_689656.2 | 6 - 7 | 776 | 109 |
GenBank mRNA | AK054835.1 | - | 4 - 5 | 564 | 109 |
AK315462.1 | - | 6 - 7 | 776 | 109 | |
BC025724.1 | - | 4 - 5 | 622 | 109 | |
KU178865.1 | - | 4 - 5 | 452 | 109 |