The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152443.2 | NP_689656.2 | 8 - 9 | 1170 | 157 |
GenBank mRNA | AK054835.1 | - | 6 - 7 | 958 | 157 |
AK315462.1 | - | 8 - 9 | 1170 | 157 | |
BC025724.1 | - | 6 - 7 | 1016 | 157 | |
KU178865.1 | - | 6 - 7 | 846 | 157 | |
KU178866.1 | - | 5 - 6 | 636 | 157 |