The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201573.1 | NP_001188502.1 | 2 - 3 | 205 | 108 |
NM_025152.2 | NP_079428.2 | 4 - 5 | 441 | 108 | |
NR_120408.1 | 4 - 5 | 441 | 108 | ||
XM_011537183.2 | XP_011535485.1 | 4 - 5 | 427 | 108 | |
XM_017021664.1 | XP_016877153.1 | 4 - 5 | 427 | 108 | |
XM_017021665.1 | XP_016877154.1 | 4 - 5 | 427 | 108 | |
XM_017021666.1 | XP_016877155.1 | 4 - 5 | 427 | 108 | |
GenBank mRNA | AK022722.1 | - | 4 - 5 | 354 | 108 |
AK295326.1 | - | 4 - 5 | 402 | 108 | |
AK301447.1 | - | 2 - 3 | 205 | 108 | |
AK316445.1 | - | 4 - 5 | 431 | 108 | |
BC024919.1 | - | 4 - 5 | 367 | 108 | |
BX248028.1 | - | 4 - 5 | 428 | 108 | |
KJ534907.1 | - | 4 - 5 | 386 | 108 | |
KJ534908.1 | - | 4 - 5 | 386 | 108 | |
KJ534909.1 | - | 4 - 5 | 386 | 108 |