The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310332.1 | NP_001297261.1 | 2 - 3 | 181 | 69 |
NM_017999.4 | NP_060469.4 | 2 - 3 | 589 | 69 | |
GenBank mRNA | AB265810.1 | - | 2 - 3 | 340 | 69 |
AK122711.1 | - | 1 - 2 | 42 | 69 | |
AK225832.1 | - | 2 - 3 | 539 | 69 | |
AK291247.1 | - | 2 - 3 | 541 | 69 | |
AY256461.1 | - | 2 - 3 | 169 | 69 | |
BC012077.1 | - | 2 - 3 | 509 | 69 | |
DA462121.1 | - | 2 - 3 | 181 | 69 |