The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310332.1 | NP_001297261.1 | 7 - 8 | 1109 | 99 |
NM_017999.4 | NP_060469.4 | 7 - 8 | 1453 | 99 | |
GenBank mRNA | AB265810.1 | - | 7 - 8 | 1204 | 99 |
AK055542.1 | - | 1 - 2 | 250 | 99 | |
AK122711.1 | - | 6 - 7 | 906 | 99 | |
AK225832.1 | - | 7 - 8 | 1403 | 99 | |
AK291247.1 | - | 7 - 8 | 1405 | 99 | |
AY256461.1 | - | 7 - 8 | 1097 | 99 | |
BC012077.1 | - | 7 - 8 | 1373 | 99 |