The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310332.1 | NP_001297261.1 | 8 - 9 | 1390 | 82 |
NM_017999.4 | NP_060469.4 | 8 - 9 | 1734 | 82 | |
GenBank mRNA | AB265810.1 | - | 8 - 9 | 1485 | 82 |
AK055542.1 | - | 2 - 3 | 531 | 82 | |
AK225832.1 | - | 8 - 9 | 1684 | 82 | |
AK291247.1 | - | 8 - 9 | 1686 | 82 | |
AY256461.1 | - | 8 - 9 | 1378 | 82 | |
BC009821.2 | - | 1 - 2 | 88 | 82 | |
BC012077.1 | - | 8 - 9 | 1654 | 82 | |
BC017376.2 | - | 1 - 2 | 78 | 82 |