The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039619.2 | NP_001034708.1 | 16 - 17 | 2073 | 77 |
NM_001282953.1 | NP_001269882.1 | 15 - 16 | 1941 | 77 | |
NM_001282954.1 | NP_001269883.1 | 15 - 16 | 1954 | 77 | |
NM_001282955.1 | NP_001269884.1 | 15 - 16 | 1856 | 77 | |
NM_001282956.1 | NP_001269885.1 | 12 - 13 | 1611 | 77 | |
NM_006109.4 | NP_006100.2 | 16 - 17 | 1988 | 77 | |
XM_017020927.1 | XP_016876416.1 | 15 - 16 | 1712 | 77 | |
GenBank mRNA | AF015913.1 | - | 16 - 17 | 1764 | 77 |
AF167572.1 | - | 16 - 17 | 1855 | 77 | |
AK075251.1 | - | 16 - 17 | 1866 | 77 | |
AK300863.1 | - | 15 - 16 | 1738 | 77 | |
AK301812.1 | - | 15 - 16 | 1747 | 77 | |
AK302240.1 | - | 15 - 16 | 1636 | 77 | |
AK315500.1 | - | 16 - 17 | 1804 | 77 | |
AL833612.1 | - | 16 - 17 | 1773 | 77 | |
BC025979.2 | - | 16 - 17 | 1776 | 77 | |
CR456741.1 | - | 16 - 17 | 1764 | 77 |