The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326358.1 | NP_001313287.1 | 13 - 14 | 2452 | 90 |
NM_199355.3 | NP_955387.1 | 13 - 14 | 2448 | 90 | |
XM_011522923.1 | XP_011521225.1 | 11 - 12 | 1859 | 90 | |
XM_011522924.1 | XP_011521226.1 | 11 - 12 | 1855 | 90 | |
XM_017022988.1 | XP_016878477.1 | 10 - 11 | 1695 | 90 | |
XM_017022989.1 | XP_016878478.1 | 10 - 11 | 1492 | 90 | |
GenBank mRNA | AJ311903.1 | - | 13 - 14 | 2108 | 90 |
AK131403.1 | - | 12 - 13 | 1955 | 90 | |
BC063283.1 | - | 13 - 14 | 2448 | 90 |